missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGD1 Antibody (OTI4G9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74519CL1
This item is not returnable.
View return policy
Description
TIGD1 Monoclonal antibody specifically detects TIGD1 in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| TIGD1 | |
| Monoclonal | |
| CoraFluor 1 | |
| EEYORE, tigger transposable element derived 1, tigger transposable element-derived protein 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-267 of human TIGD1(NP_663748) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI4G9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 200765 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction