missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141382
This item is not returnable.
View return policy
Description
Sequence homology: Chimp (93%).
This gene encodes a member of the kelch family of proteins, which are characterized by kelch repeat motifs and a POZ/BTB protein-binding domain. It is thought that kelch repeats are actin binding domains. However, the specific function of this protein has not been determined. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| TIGAR | |
| Polyclonal | |
| Unconjugated | |
| TIGAR | |
| 9630033F20Rik; AA793651; AI595337; C12orf5; C27H12orf5; C79710; C85509; FR2BP; Fructose-2,6-bisphosphatase TIGAR; fructose-2,6-bisphosphate 2-phosphatase; hypothetical protein LOC502894; LOC502894; probable fructose-2,6-bisphosphatase TIGAR; TIGAR; TP53 induced glycolysis regulatory phosphatase; TP53-induced glycolysis and apoptosis regulator; TP53-induced glycolysis regulatory phosphatase; transactivated by NS3TP2 protein; Trp53 induced glycolysis repulatory phosphatase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 319801, 502894, 57103 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| Q8BZA9, Q9NQ88 | |
| TIGAR | |
| A portion of amino acids 140-190 of human TIGAR. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction