missing translation for 'onlineSavingsMsg'
Learn More

TIGAR/C12orf5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-24754SS

 View more versions of this product

Product Code. 18149001

  • £167.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIGAR/C12orf5
Polyclonal
Unconjugated
0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide
chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
Rabbit
Protein G purified
RUO
Primary
The amino acid sequence used as immunogen for this antibody is 100% homologous in human and 93% homologous in chimp.
Store at -20C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1-4 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Q9NQ88
TIGAR
A portion of amino acids 140-190 of human TIGAR was used as the immunogen for the antibody.
0.025 mg
Lipid and Metabolism
57103
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only