missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGAR/C12orf5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £367.00
Specifications
| Antigen | TIGAR/C12orf5 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18286912
|
Novus Biologicals
NBP1-49534SS |
0.025 mL |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18748213
|
Novus Biologicals
NBP1-49534 |
£367.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TIGAR/C12orf5 Polyclonal specifically detects TIGAR/C12orf5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TIGAR/C12orf5 | |
| Western Blot 1:5000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein | |
| TIGAR | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Q9NQ88, Q8BZA9 | |
| 57103 | |
| A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title