Learn More
Invitrogen™ TIF1 gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580187
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human K562 whole cell, human Jurkat whole cell, rat brain tissue, mouse brain tissue. IHC: human lung adenocarcinoma tissue, human ovarian serous adenocarcinoma tissue. ICC/IF: A431 cell. Flow: U87 cell, A431 cell.
TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.
Specifications
| TIF1 gamma | |
| Polyclonal | |
| Unconjugated | |
| TRIM33 | |
| 8030451N04Rik; AI413936; E3 ubiquitin-protein ligase TRIM33; Ecto; Ectodermin homolog; Kiaa1113; Protein Rfg7; PTC7; RET-fused gene 7 protein; RFG7; RING-type E3 ubiquitin transferase TRIM33; TF1G; TIF 1-g; Tif1g; TIF1GAMMA; TIF1-gamma; TIFGAMMA; Transcription intermediary factor 1-gamma; transcriptional intermediary factor 1 gamma; Trim33; tripartite motif containing 33; tripartite motif protein 33; tripartite motif-containing 33; tripartite motif-containing protein 33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 365894, 51592, 94093 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q99PP7, Q9UPN9 | |
| TRIM33 | |
| E.coli-derived human TIF1 gamma recombinant protein (Position: M1001-K1127). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.