missing translation for 'onlineSavingsMsg'
Learn More

TIF1 gamma Antibody, mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18401598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401598 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401598 Supplier Novus Biologicals Supplier No. NBP281762MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIF1 gamma Polyclonal antibody specifically detects TIF1 gamma in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIF1 gamma
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias E3 ubiquitin-protein ligase TRIM33, EC 6.3.2, EC 6.3.2.-, Ectodermin homolog, KIAA1113, Protein Rfg7, PTC7FLJ32925, RET-fused gene 7 protein, RFG7FLJ11429, TF1G, TIF1-gamma, TIF1GAMMA, TIF1GECTO, TIFGAMMA, Transcription intermediary factor 1-gamma, transcriptional intermediary factor 1 gamma, tripartite motif containing 33, tripartite motif-containing 33, Tripartite motif-containing protein 33
Host Species Rabbit
Immunogen TIF1 gamma antibody was raised against an 18 amino acid peptide near the center of human TIF1 gamma. The immunogen is located within amino acids 580 - 630 of TIF1 gamma. Amino Acid Squence: AQNAARIPGIPRHSGPQY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51592
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.