missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIF1 gamma Antibody (6F4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00051592-M02
This item is not returnable.
View return policy
Description
TIF1 gamma Monoclonal antibody specifically detects TIF1 gamma in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| TIF1 gamma | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| E3 ubiquitin-protein ligase TRIM33, EC 6.3.2, EC 6.3.2.-, Ectodermin homolog, KIAA1113, Protein Rfg7, PTC7FLJ32925, RET-fused gene 7 protein, RFG7FLJ11429, TF1G, TIF1-gamma, TIF1GAMMA, TIF1GECTO, TIFGAMMA, Transcription intermediary factor 1-gamma, transcriptional intermediary factor 1 gamma, tripartite motif containing 33, tripartite motif-containing 33, Tripartite motif-containing protein 33 | |
| TRIM33 (NP_056990, 1006 a.a. ~ 1105 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VKKKLQKKHSQHYQIPDDFVADVRLIFKNCERFNEMMKVVQVYADTQEINLKADSEVAQAGKAVALYFEDKLTEIYSDRTFAPLPEFEQEEDDGEVTEDS | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 6F4 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| NP_056990 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 51592 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction