missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIF1 alpha Mouse anti-Human, Clone: PCRP-TRIM24-1B12, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07464-100ug
This item is not returnable.
View return policy
Description
TIF1 alpha Monoclonal specifically detects TIF1 alpha in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
Specifications
| TIF1 alpha | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| E3 ubiquitin-protein ligase TRIM24, EC 6.3.2, EC 6.3.2.-, hTIF1, PTC6, RING finger protein 82, RNF82Tif1a, TIF1-alpha, TIF1ATIF1ALPHA, TIF1TF1A, transcription intermediary factor 1-alpha, transcriptional intermediary factor 1, tripartite motif containing 24, tripartite motif-containing 24, Tripartite motif-containing protein 24 | |
| Recombinant full-length human TIF1 alpha protein | |
| 100 μg | |
| Protein Kinase, Zinc Finger | |
| 8805 | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Immunofluorescence, Peptide Array | |
| PCRP-TRIM24-1B12 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction