missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Tie-1 Antibody Pair [HRP]

Product Code. 18282057 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18282057

Brand: Novus Biologicals™ NBP27947815PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Tie-1.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78 pg/mL
Assay Sensitivity 78.13-5000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Tie-1.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1
Gene ID (Entrez) 7075
Gene Symbol TIE1
Host Species Mouse
Kit Contents
  • Standard: Each vial contains 160 ng of recombinant Tie-1.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Tie-1 monoclonal antibody,
  • Detection Antibody: 0.5 mg/mL mouse anti-Tie-1 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Angiogenesis, Cancer, Protein Kinase
    Storage Requirements Storage is content dependent.
    Target Tie-1
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 5 ng/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.