missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tie-1 Antibody (6F12), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-18610DL594
This item is not returnable.
View return policy
Description
Tie-1 Monoclonal antibody specifically detects Tie-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF
Specifications
| Tie-1 | |
| Monoclonal | |
| DyLight 594 | |
| EC 2.7.10, EC 2.7.10.1, JTK14soluble TIE1 variant 3, receptor tyrosine kinase, soluble TIE1 variant 1, soluble TIE1 variant 2, soluble TIE1 variant 5, TIEsoluble TIE1 variant 4, tyrosine kinase with immunoglobulin and epidermal growth factor homologydomains 1, tyrosine kinase with immunoglobulin-like and EGF-like domains 1, tyrosine-protein kinase receptor Tie-1 | |
| Recombinant human soluble extracellular TIE-1 | |
| 0.1 mL | |
| Angiogenesis, Cancer, Protein Kinase | |
| 7075 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| 6F12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction