missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TID1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527226
This item is not returnable.
View return policy
Description
Recommended positive controls: HepG2, HCT116. Predicted reactivity: Mouse (87%), Rat (87%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TID1 is a human homolog of the Drosophila tumor suppressor lethal (2) tumorous imaginal discs, l(2)tid and encodes two mitochondrial matrix localized splice variants of human Tid-1 designated hTid-1S and hTid-1L. These proteins are the conserved members of the DnaJ family of proteins which act as cochaperones for mitochondrial Hsp70. They contain a conserved tetrahedrical J domain which binds to Hsp70 chaperones and activates their ATPase activity. Expression of hTid-1L increases apoptosis induced by DNA damaging agents as mitomycin-C and TNF-a. A J-domain mutant of hTid-1L can dominantly suppress apoptosis and in sharp contrast the J-domain mutant of hTid-1S increases apoptosis. Expression of hTid-1S and hTid-1L affects cytochrome c release from the mitochondria and caspase 3 activation, while activation of caspase 8 is unaffected. It is strongly suggested that these two splice variants exert their anti- and pro- apoptotic effects through discrete substrates and activities. Hence the relative abundance of these proteins or their substrates may allow the mitochondria to dampen or enhance the apoptotic signals.
Specifications
| TID1 | |
| Polyclonal | |
| Unconjugated | |
| DNAJA3 | |
| 1200003J13Rik; 1810053A11Rik; C81173; DnaJ (Hsp40) homolog, subfamily A, member 3; DnaJ heat shock protein family (Hsp40) member A3; dnaJ homolog subfamily A member 3, mitochondrial; DnaJ protein Tid-1; Dnaja3; HCA57; Hepatocellular carcinoma-associated antigen 57; hTID-1; mTid-1; Tid1; Tid-1; Tid1l; Tumorous imaginal discs protein Tid56 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9093 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.46 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q96EY1 | |
| DNAJA3 | |
| Recombinant protein encompassing a sequence within the center region of human TID1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction