missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TID1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38368-20ul
This item is not returnable.
View return policy
Description
TID1 Polyclonal antibody specifically detects TID1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TID1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DnaJ (Hsp40) homolog, subfamily A, member 3, DnaJ protein Tid-1, HCA57, Hepatocellular carcinoma-associated antigen 57, hTid-1FLJ45758, TID1dnaJ homolog subfamily A member 3, mitochondrial, tumorous imaginal discs homolog, Tumorous imaginal discs protein Tid56 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 221-480 of human TID1 (NP_005138.3).,, Sequence:, AKGVNKEFTVNIMDTCERCNGKGNEPGTKVQHCHYCGGSGMETINTGPFVMRSTCRRCGGRGSIIISPCVVCRGAGQAKQKKRVMIPVPAGVEDGQTVRMPVGKREIFITFRVQKSPVFRRDGADIHSDLFISIAQALLGGTARAQGLYETINVTIPPGTQTDQKIRMGGKGIPRINSYGYGDHYIHIKIRVPKRLTSRQQSLILSYAEDETDVEGTVNGVTLTSSGGSTMDSSAGSKARREAGEDEEGFLSKLKKMFTS | |
| 20 μL | |
| Apoptosis | |
| 9093 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction