missing translation for 'onlineSavingsMsg'
Learn More

TICRR Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30156983 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30156983 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30156983 Supplier Novus Biologicals Supplier No. NBP241283CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TICRR Polyclonal antibody specifically detects TICRR in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TICRR
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias C15orf42, Chromosome 15 Open Reading Frame 42, SLD3, SLD3 Homolog, SLD3 Homolog (S. Cerevisiae), TOPBP1-Interacting Checkpoint And Replication Regulator, TOPBP1-Interacting Replication-Stimulating Protein, TopBP1-Interacting, Replication-Stimulating Protein, Treslin
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the carboxy terminus of human TICRR. The immunogen is located within amino acids 1840 - 1890 of TICRR. Amino Acid Squence: FQGKTPSSQSKDPRDEDVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 90381
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.