missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ticlopidine EP Impurity C (as Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Ticlopidine EP Impurity C (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | (2-Chlorophenyl)methanamine Hydrochloride |
| CAS | 22680-44-0 |
| Analyte or Component Names | (2-Chlorophenyl)methanamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H8 Cl N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ticlopidine Hydrochloride Imp. C (EP), Ticlopidine Imp. C (EP), (2-Chlorophenyl)methanamine Hydrochloride, Ticlopidine Hydrochloride Imp. C (EP) as Hydrochloride, Ticlopidine Hydrochloride Impurity C as Hydrochloride, Ticlopidine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C7H8ClN.ClH/c8-7-4-2-1-3-6(7)5-9;/h1-4H,5,9H2;1H |
| SMILES | Cl.NCc1ccccc1Cl |
| IUPAC Name | (2-chlorophenyl)methanamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction