missing translation for 'onlineSavingsMsg'
Learn More

Melford Ticarcillin Disodium Salt

Product Code. 30018857
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 30018857

Brand: Melford T365005.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ticarcillin disodium salt is an antibiotic that belongs to the class of drugs known as penicillin.

Ticarcillin disodium salt is an antibiotic that belongs to the class of drugs known as penicillin. It is a broad-spectrum antibiotic effective against various bacterial infections.

Ticarcillin disodium salt is commonly used in the field of life sciences for research applications.

In the life science marketplace, ticarcillin disodium salt is primarily utilized as a selective agent in microbiology and molecular biology experiments.

It is often incorporated into culture media to inhibit the growth of certain bacteria, allowing for the selective growth of desired organisms or the detection of specific bacterial strains.

Researchers and scientists working in fields such as microbiology, immunology, and molecular biology may employ ticarcillin disodium salt to study bacterial resistance mechanisms, evaluate the efficacy of antimicrobial compounds, or isolate and identify specific bacterial species.

Specifications

Chemical Name or Material Ticarcillin Disodium Salt
CAS 4697-14-7
Quantity 5 g
Molecular Formula C15H14N2Na2O6S2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.