missing translation for 'onlineSavingsMsg'
Learn More

Ticarcillin disodium and clavulanate potassium (15:1)

Código de producto. 15490617
Cambiar vista
Click to view available options
Quantity:
10 g
Packaging:
Plastic bottle
Tamaño de la unidad:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging Tamaño de la unidad
15490617 10 g Plastic bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15490617 Proveedor Thermo Scientific Alfa Aesar N.º de proveedor J66659.09
  • ONLINE EXCLUSIVE PRICE
    £148.92
    £120.65 / 10g
    Save £28.27
    18% Off
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 86482-18-0 | C23H25N3O11S2 | 583.583 g/mol

Combination provides greater efficacy against beta--lactam resistant strains

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

CAS 86482-18-0
Physical Form Powder
Quantity 10 g
Packaging Plastic bottle
Molecular Formula C23H25N3O11S2
MDL Number MFCD26967928
Synonym ticarcillin-clavulanic acid, clavulanic acid mixture with ticarcillin, clavulanic acid-ticarcillin mixt., ticarcillin-clavulanic acid mixt., ticarcillin disodium and clavulanate potassium, timentin in plastic container, 4-oxa-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 3-2-hydroxyethylidene-7-oxo-, 2r,3z,5r-, mixt. with 2s,5r,6r-6-2r-carboxy-3-thienylacetyl amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 4-oxa-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, 3-2-hydroxyethylidene-7-oxo-, 2r-2-alpha,3z,5-alpha-, mixt. with 2s-2-alpha,5-alpha,6-beta s*-6-carboxy-3-thienylacetyl amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid, clavulanic acid-ticarcillin mixt
Solubility Information Freely soluble in water
InChI Key XWMVMWTVLSLJGY-FAJPTIRJSA-N
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CSC=C3)C(=O)O)C(=O)O)C.C1C2N(C1=O)C(C(=CCO)O2)C(=O)O
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-carboxy-2-thiophen-3-ylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;(2R,3Z,5R)-3-(2-hydroxyethylidene)-7-oxo-4-oxa-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Molecular Weight (g/mol) 583.583
PubChem CID 6437075
Chemical Name or Material Ticarcillin disodium and clavulanate potassium (15:1)
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Ticarcillin disodium with clavulanate potassium (15:1)
Palabra de advertencia
  • Danger
Clasificación
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Otros peligros
  • MIXTURE LIST-Contain: Ticarcillin disodiumsalt, clavulanate potassium

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.