missing translation for 'onlineSavingsMsg'
Learn More

TICAM2 Polyclonal Antibody

Product Code. 12858212
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12858212 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12858212 Supplier Invitrogen Supplier No. PA520037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TICAM2 Polyclonal Antibody

TICAM2 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen TICAM2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TICAM2
Gene Accession No. Q86XR7
Gene Alias B430113A10; cytoplasmic adaptor; MyD88-4; NF-kappa-B-activating protein 502; putative NF-kappa-B-activating protein 502; TAG; TICAM2; TICAM-2; TIR domain-containing adapter molecule 2; TIR domain-containing adapter protein; TIRAP3; Tirp; toll like receptor adaptor molecule 2; toll/interleukin-1 receptor (TIR) domain-containing adapter protein; toll/interleukin-1 receptor domain-containing protein; Toll-interleukin I receptor domain (TIR)-containing adaptor molecule (TICAM) 2; toll-like receptor adaptor molecule 2; toll-like receptor adaptor protein 3; Tram; Trif; TRIF-related adapter molecule; TRIF-related adapter molecule TRAM; TRIF-related adaptor molecule
Gene Symbols TICAM2
Host Species Rabbit
Immunogen A peptide corresponding to amino acids at the C-terminus of human TIRP.
Purification Method Ion-exchange chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353376
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.