missing translation for 'onlineSavingsMsg'
Learn More

Tibenelast sodium, MedChemExpress

Product Code. 16486237
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486237 5 mg 5mg
16433518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16486237 Supplier Medchem Express Supplier No. HY101705
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tibenelast sodium is a phosphodiesterase inhibitor.

Tibenelast sodium is a phosphodiesterase inhibitor.

Specifications

Chemical Name or Material Tibenelast sodium
CAS 105102-18-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C13H13NaO4S
Quantity 5 mg
Synonym LY 186655
Solubility Information DMSO : 50 mg/mL (173.44 mM; Need ultrasonic)
SMILES O=C(C1=CC2=CC(OCC)=C(OCC)C=C2S1)[O-].[Na+]
Molecular Weight (g/mol) 288.29
Formula Weight 288.29
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.