missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiaprofenic Acid Ethyl Ester, Mikromol™
Description
Tiaprofenic Acid Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Tiaprofenic Acid Ethyl Ester |
| CAS | 127266-13-1 |
| Analyte or Component Names | Tiaprofenic Acid Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H16 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Tiaprofenic Acid Ethyl Ester, 5-Benzoyl-alpha-methyl-2-thiopheneacetic acid ethyl ester |
| InChI Formula | InChI=1S/C16H16O3S/c1-3-19-16(18)11(2)13-9-10-14(20-13)15(17)12-7-5-4-6-8-12/h4-11H,3H2,1-2H3 |
| SMILES | CCOC(=O)C(C)c1ccc(s1)C(=O)c2ccccc2 |
| IUPAC Name | ethyl 2-(5-benzoylthiophen-2-yl)propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?