missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiapride hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Tiapride hydrochloride is a selective and orally active D2 and D3 dopamine receptors antagonist with IC50 values of 110-320 nM and 180 nM, respectively. Tiapride hydrochloride shows anti-dyskinetic activity and anxiolytic activity. Tiapride hydrochloride is a neuroleptic agent.
Specifications
Specifications
| Chemical Name or Material | Tiapride hydrochloride |
| CAS | 51012-33-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H25ClN2O4S |
| Quantity | 100 mg |
| Solubility Information | H2O : ≥ 200 mg/mL (548.11 mM) ∣DMSO : 31.25 mg/mL (85.64 mM; Need ultrasonic) |
| SMILES | O=C(NCCN(CC)CC)C1=CC(S(=O)(C)=O)=CC=C1OC.[H]Cl |
| Molecular Weight (g/mol) | 364.89 |
| Formula Weight | 364.89 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction