missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiapride hydrochloride, Dr. Ehrenstorfer
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Tiapride hydrochloride |
| CAS | 51012-33-0 |
| Analyte or Component Names | Tiapride hydrochloride |
| Shipping Condition | +20°C |
| Molecular Formula | C15 H24 N2 O4 S . Cl H |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | N-[2-(Diethylamino)ethyl]-2-methoxy-5-(methylsulfonyl)-benzamide monohydrochloride, Gramalil, Italprid, Luxoben, Sereprile, Tiapridal, Tiapride hydrochloride, Tiapridex |
| InChI Formula | InChI=1S/C15H24N2O4S.ClH/c1-5-17(6-2)10-9-16-15(18)13-11-12(22(4,19)20)7-8-14(13)21-3;/h7-8,11H,5-6,9-10H2,1-4H3,(H,16,18);1H |
| SMILES | Cl.CCN(CC)CCNC(=O)c1cc(ccc1OC)S(=O)(=O)C |
| IUPAC Name | N-[2-(diethylamino)ethyl]-2-methoxy-5-methylsulfonylbenzamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction