missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tianeptine-d4 (Major), TRC
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | Tianeptine-d4 |
| Molecular Formula | C21H21D4ClN2O4S |
| Quantity | 0.5 mg |
| Synonym | 7-[(3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2]thiazepin-11-yl)amino]heptanoic Acid-d4, S,S-Dioxide, (+/-)-7-[(3-Chloro-6,11-dihydro-6-methyldibenzo[c,f][1,2] thiazepin-11-yl)amino]heptanoic Acid-d4 S,S-Dioxide, Coaxil-d4 |
| InChI Formula | InChI=1S/C21H25ClN2O4S/c1-24-18-9-6-5-8-16(18)21(23-13-7-3-2-4-10-20(25)26)17-12-11-15(22)14-19(17)29(24,27)28/h5-6,8-9,11-12,14,21,23H,2-4,7,10,13H2,1H3,(H,25,26)/i3D2,7D2 |
| SMILES | ClC1=CC=C(C(NCC([2H])([2H])C([2H])([2H])CCCC(O)=O)C(C=CC=C2)=C2N(C)S3(=O)=O)C3=C1 |
| Molecular Weight (g/mol) | 440.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction