missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiamulin fumarate, MedChemExpress
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
Tiamulin fumarate (Thiamutilin fumarate) is a diterpenic compound that widely used in swine for the control of infectious diseases, including swine dysentery and enzootic pneumonia.
Specifications
Specifications
| Chemical Name or Material | Tiamulin fumarate |
| CAS | 55297-96-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C32H51NO8S |
| Quantity | 250 mg |
| Synonym | Thiamutilin fumarate |
| Solubility Information | DMSO : ≥ 100 mg/mL (163.99 mM) ∣H2O : 100 mg/mL (163.99 mM; Need ultrasonic) |
| SMILES | O=C(O[C@H]1[C@]([C@H](C)CC2)(C)[C@@](C(CC3)=O)([H])[C@]32[C@@H](C)[C@H](O)[C@](C)(C=C)C1)CSCCN(CC)CC.O=C(O)/C=C/C(O)=O |
| Molecular Weight (g/mol) | 609.81 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction