missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIAM2 Recombinant Rabbit Monoclonal Antibody (K01_2J29)
Description
TIAM2 Recombinant Monoclonal Antibody for Western Blot
Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP- dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Mediates extracellular laminin signals to activate Rac1, contributing to neurite growth. Involved in lamellipodial formation and advancement of the growth cone of embryonic hippocampal neurons. Promotes migration of neurons in the cerebral cortex. When overexpressed, induces membrane ruffling accompanied by the accumulation of actin filaments along the altered plasma membrane. Activates specifically RAC1, but not CDC42 and RHOA.
Specifications
Specifications
| Antigen | TIAM2 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2J29 |
| Concentration | 1.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gene | Tiam2 |
| Gene Accession No. | Q8IVF5 |
| Gene Alias | 3000002F19Rik; KIAA2016; mKIAA2016; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; SIF and TIAM1-like exchange factor; Stef; T cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; T-cell lymphoma invasion and metastasis 2; RAS-related C3 botulinum substrate 1, guanine nucleotide exchange factor 1; TIAM2; TIAM-2; T-lymphoma invasion and metastasis-inducing protein 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?