missing translation for 'onlineSavingsMsg'
Learn More

TIAM2 Antibody, Novus Biologicals™

Product Code. 18357969 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357969 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357969 Supplier Novus Biologicals Supplier No. H00026230B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TIAM2 Polyclonal antibody specifically detects TIAM2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAM2
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_001010927
Gene Alias FLJ41865, KIAA2016, SIF and TIAM1-like exchange factor, STEFTIAM-2, T-cell lymphoma invasion and metastasis 2, T-lymphoma invasion and metastasis-inducing protein 2
Host Species Mouse
Immunogen TIAM2 (NP_001010927, 1 a.a. - 626 a.a.) full-length human protein. MEGPRENQDPPPRPLARHLSDADRLRKVIQELVDTEKSYVKDLSCLFELYLEPLQNETFLTQDEMESLFGSLPEMLEFQKVFLETLEDGISASSDFNTLETPSQFRKLLFSLGGSFLYYADHFKLYSGFCANHIKVQKVLERAKTDKAFKAFLDARNPTKQHSSTLESYLIKPVQRVLKYPLLLKELVSLTDQESEEHYHLTEALKAMEKVASHINEMQKIYEDYGTVFDQLVAEQSGTEKEVTELSMGELLMHSTVSWLNPFLSLGKARKDLELTVFVFKRAVILVYKENCKLKKKLPSNSRPAHNSTDLDPFKFRWLIPISALQVRLGNPAGTENNSIWELIHTKSEIEGRPETIFQLCCSDSESKTNIVKVIRSILRENFRRHIKCELPLEKTCKDRLVPLKNRVPVSAKLASSRSLKVLKNSSSNEWTGETGKGTLLDSDEGSLSSGTQSSGCPTAEGRQDSKSTSPGKYPHPGLADFADNLIKESDILSDEDDDHRQTVKQGSPTKDIEIQFQRLRISEDPDVHPEAEQQPGPESGEGQKGGEQPKLVRGHFCPIKRKANSTKRDRGTLLKAQIRHQSLDSQSENATIDLNSVLEREFSVQSLTSVVSEECFYETESHGKS
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26230
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.