missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIAM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-66594
This item is not returnable.
View return policy
Description
TIAM1 Polyclonal specifically detects TIAM1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| TIAM1 | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10 - 1:500 | |
| FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1 | |
| Rabbit | |
| Affinity Purified | |
| Cancer | |
| 7074 | |
| Human, Mouse | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS, pH 7.2 with 0.09% Sodium Azide | |
| TIAM1 | |
| Human Tiam1 (catalytic domain/ C-terminal domain) is an affinity purified rabbit polyclonal antiserum derived by injection of rabbits with a GST fusion protein with the C-terminal amino acids 904-1511 of Tiam 1. The antiserum was purified by affinity chro | |
| 0.1 mL | |
| Primary | |
| Human Tiam1 (catalytic domain/ C-terminal domain) recognizes the DH-PH regions of Tiam1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction