missing translation for 'onlineSavingsMsg'
Learn More

TIAM1 Antibody, Novus Biologicals™

Product Code. 18363961 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18363961 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18363961 Supplier Novus Biologicals Supplier No. NBP275096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

TIAM1 Polyclonal specifically detects TIAM1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIAM1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Peptide ELISA
Formulation TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
Gene Alias FLJ36302, human T-lymphoma invasion and metastasis inducing TIAM1 protein, 10TIAM-1, T-cell lymphoma invasion and metastasis 1, T-lymphoma invasion and metastasis-inducing protein 1
Gene Symbols TIAM1
Host Species Goat
Immunogen Peptide with sequence C-EDFAPSRKLNTEI, from the C Terminus of the protein sequence according to NP_003244; NP_001340613.1; NP_001340614.1; NP_001340615.1; NP_001340616.1.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7074.0
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.