missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tiagabine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Tiagabine Hydrochloride |
| CAS | 145821-59-6 |
| Analyte or Component Names | Tiagabine Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C20 H25 N O2 S2 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Tiagabine Hydrochloride, (3R)-1-[4,4-Bis(3-methyl-2-thienyl)-3-butenyl]-3-piperidinecarboxylic acid hydrochloride, (R)-1-[4,4-Bis(3-methyl-2-thienyl)-3-butenyl]-3-piperidinecarboxylic acid hydrochloride, ABT 569, Abbott 70569 hydrochloride, Abbott 70569.1, Gabitril, NNC 05-0328, NO 05-0328, Tiabex, Tiagabine hydrochloride |
| InChI Formula | InChI=1S/C20H25NO2S2.ClH/c1-14-7-11-24-18(14)17(19-15(2)8-12-25-19)6-4-10-21-9-3-5-16(13-21)20(22)23;/h6-8,11-12,16H,3-5,9-10,13H2,1-2H3,(H,22,23);1H/t16-;/m1./s1 |
| Solution Type | Single Solution |
| SMILES | Cl.Cc1ccsc1C(=CCCN2CCC[C@H](C2)C(=O)O)c3sccc3C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction