missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562325
This item is not returnable.
View return policy
Description
Immunogen sequence: MSSPSSPFRE QSFLCAAGDA GEESRVQVLK NEVRRGSPVL LGWVEQAYAD KCV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 28%.
TIAF-1 (TGFB1-induced anti-apoptotic factor 1), also known as MYO18A, is a TGFB1-induced anti-apoptotic factor that functions to inhibit the cytotoxic effects of TNF-alpha, TRADD and FADD. It localizes to the nucleus and significantly up-regulated in activated helper T lymphocytes (TH2) in chronic kidney and liver allograft rejection. Suppression of TIAF1 expression by siRNA prevents UV irradiation-mediated p53 phosphorylation and nuclear translocation. TIAF1 is essential in the p53- and WOX1-mediated cell death and shows an aggregation-dependent control of tumor progression and metastasis, and regulation of cell death.
Specifications
| TIAF1 | |
| Polyclonal | |
| Unconjugated | |
| TIAF1 | |
| 12 kDa TGF-beta-1-induced antiapoptotic factor; MAJN; molecule associated with Jak-3 N-terminal; MYO18A; MYSPDZ; SPR210; TGFB1-induced anti-apoptotic factor 1; TGF-beta-1-induced antiapoptotic factor 1; TIAF; TIAF1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9220 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95411 | |
| TIAF1 | |
| Recombinant protein corresponding to Human TIAF1. Recombinant protein control fragment (Product #RP-100356). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction