missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TIAF1 Polyclonal antibody specifically detects TIAF1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry
Specifications
Specifications
| Antigen | TIAF1 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunocytochemistry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 12 kDa TGF-beta-1-induced antiapoptotic factor, KIAA0216, MAJN, molecule associated with Jak-3 N-terminal, MYO18A, MYSPDZ, SPR210, TGFB1-induced apoptotic factor 1, TGF-beta-1-induced antiapoptotic factor 1 |
| Host Species | Rabbit |
| Immunogen | TIAF1 antibody was raised against a 16 amino acid peptide near the amino terminus of human TIAF1 . The immunogen is located within the first 50 amino acids of TIAF1. Amino Acid Squence: CAGEESRVQVLKNEVRRG |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?