missing translation for 'onlineSavingsMsg'
Learn More

TIA1 Antibody, Novus Biologicals™

Product Code. 30228695 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30228695 20 μL 20µL
30227552 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30228695 Supplier Novus Biologicals Supplier No. NBP33524420ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIA1 Polyclonal antibody specifically detects TIA1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIA1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias cytotoxic granule-associated RNA-binding protein, nucleolysin TIA-1 isoform p40, p40-TIA-1, p40-TIA-1 (containing p15-TIA-1), RNA-binding protein TIA-1, T-cell-restricted intracellular antigen-1, TIA-1, TIA1 cytotoxic granule-associated RNA binding protein, TIA1 cytotoxic granule-associated RNA-binding protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 287-386 of human TIA1 (NP_071505.2).,, Sequence:, MEDEMPKTLYVGNLSRDVTEALILQLFSQIGPCKNCKMIMDTAGNDPYCFVEFHEHRHAAAALAAMNGRKIMGKEVKVNWATTPSSQKKDTSSSTVVSTQRSQDHFHVFVGDLSPEITTEDIKAAFAPFGRISDARVVKDMATGKSKGYGFVSFFNKWDAENAIQQMGGQWLGGRQIRTNWATRKPPAPKSTY
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 7072
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.