missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THZ1-R, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
THZ1-R is a non-cysteine reactive analog of THZ1 which displays diminished activity for CDK7 inhibition. THZ1-R binds to CDK7 with a Kd of 142 nM.
Specifications
Specifications
| Chemical Name or Material | THZ1-R |
| Physical Form | Powder |
| CAS | 1621523-07-6 |
| Color | Light Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C31H30ClN7O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 100 mg/mL (176.03 mM) |
| SMILES | ClC1=CN=C(NC2=CC(NC(C3=CC=C(NC(CCCN(C)C)=O)C=C3)=O)=CC=C2)N=C1C4=CNC5=CC=CC=C54 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction