missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THZ1 Hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
THZ1 Hydrochloride is a selective and potent covalent CDK7 inhibitor with an IC50 of 3.2 nM. THZ1 Hydrochloride also inhibits the closely related kinases CDK12 and CDK13 and downregulates MYC expression.
Specifications
Specifications
| Chemical Name or Material | THZ1 Hydrochloride |
| Physical Form | Solid |
| Color | Yellow |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C31H29Cl2N7O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : 22.5 mg/mL (37.34 mM; Need ultrasonic and warming) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| SMILES | ClC1=CN=C(NC2=CC(NC(C3=CC=C(NC(/C=C/CN(C)C)=O)C=C3)=O)=CC=C2)N=C1C4=CNC5=CC=CC=C54.Cl |
| Molecular Weight (g/mol) | 602.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction