missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine sulfate, MedChemExpress

Product Code. 16473348
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473348 10 mg 10mg
16456077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16473348 Supplier Medchem Express Supplier No. HY101406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Thyroxine sulfate is a thyroid hormone metabolite.

Thyroxine sulfate is a thyroid hormone metabolite.

Specifications

Chemical Name or Material Thyroxine sulfate
CAS 77074-49-8
Color White
Purity Grade Notes Research
Molecular Formula C15H11I4NO7S
Quantity 10 mg
Synonym T4 Sulfate
Solubility Information DMSO : 140 mg/mL (163.37 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC(I)=C(C(I)=C1)OC2=CC(I)=C(OS(=O)(O)=O)C(I)=C2)C(O)=O
Molecular Weight (g/mol) 856.93
Formula Weight 856.93
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.