missing translation for 'onlineSavingsMsg'
Learn More

Thyroxine Antibody (C4), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-43289

Product Code. 30279635

  • £237.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Thyroxine Monoclonal antibody specifically detects Thyroxine in Nonspecies specific samples. It is validated for ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thyroxine
Monoclonal
1 mg/mL
ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunohistochemistry-Frozen 5-20 μg/mL
T4
BSA - Thyroxine
100 μL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2b κ
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
C4
Unconjugated
0.01 M PBS, pH 7.4, 50% glycerol
Mouse
Protein A or G purified
RUO
Non-species specific
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.