missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thyroid Stimulating Hormone Monoclonal Antibody (P2D9)

Product Code. 12808062
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12808062 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12808062 Supplier Invitrogen™ Supplier No. MIT0412

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIT0412 targets Thyroid Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIT0412 immunogen is human Thyroid Stimulating Hormone. MIT0412 detects Thyroid Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa. MIT0412 was formerly sold as a Seradyn product.

The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroid Stimulating Hormone
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone P2D9
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TSHB
Gene Accession No. P01215, P01222
Gene Alias aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; Cga; Cga1; CG-alpha; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin alpha chain; FSHA; FSH-alpha; Glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; LHA; LSH-alpha; luteinizing hormone alpha chain; Lutropin alpha chain; pituitary hormone alpha subunit; thyroid stimulating hormone beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone alpha chain; Thyroid-stimulating hormone subunit beta; Thyrotropin; Thyrotropin alfa; Thyrotropin alpha chain; thyrotropin beta chain; Thyrotropin subunit beta; Tsha; TSH-alpha; Tshb; TSH-B; TSH-beta
Gene Symbols CGA, TSHB
Host Species Mouse
Immunogen Human Thyroid Stimulating Hormone.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081, 7252
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.