Learn More
Invitrogen™ Thyroid Stimulating Hormone Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHTSHX5
Additional Details : Weight : 0.33820kg
Description
The Human TSH (CGA) ELISA quantitates Hu TSH in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TSH. Principle of the method The Human TSH solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Thyroid-stimulating hormone (TSH), also known as thyrotropin, is secreted from cells in the anterior pituitary called thyrotrophs, finds its receptors on epithelial cells in the thyroid gland, and stimulates that gland to synthesize and release thyroid hormones. TSH is a glycoprotein hormone composed of two subunits which are non-covalently bound to one another.Specifications
P01215 | |
4 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
1081 | |
<12% | |
SAV-HRP, Adhesive Plate Covers, Standard, Wash Buffer, Stop Solution, Pre-coated 96 well plates, Assay Diluent Concentrate, Biotinylated Detection Antibody, Chromogen | |
5 x 96 Tests | |
Endocrinology | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
2.744 pg/mL to 2000 pg/mL | |
4 pg/mL to 2000 pg/mL | |
Biotin | |
Hormones & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF. | |
Human | |
Colorimetric Microplate Reader | |
CG-ALPHA,FSHA,GPHA1,GPHa,HCG,LHA,TSHA | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
RP11-156F23.1, CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA | |
1 hr. 20 min. |