missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid receptor-interacting protein 13 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04046
This item is not returnable.
View return policy
Description
Thyroid receptor-interacting protein 13 Polyclonal specifically detects Thyroid receptor-interacting protein 13 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Thyroid receptor-interacting protein 13 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| 16E1BP, 16E1-BP, HPV16 E1 protein binding protein, Human papillomavirus type 16 E1 protein-binding protein, thyroid hormone receptor interactor 13HPV16 E1 protein-binding protein, thyroid receptor interacting protein 13, thyroid receptor-interacting protein 13, TR-interacting protein 13, TRIP-13 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
| NP_004228.1 | |
| TRIP13 | |
| The immunogen recognized by this antibody maps to a region between residue 382 and 432 of human Thyroid Hormone Receptor Interactor 13 using the numbering given in entry NP_004228.1 (GeneID 9319). | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 9319 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction