missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid receptor-interacting protein 13 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04045C
This item is not returnable.
View return policy
Description
Thyroid receptor-interacting protein 13 Polyclonal antibody specifically detects Thyroid receptor-interacting protein 13 in Human samples. It is validated for Western Blot
Specifications
| Thyroid receptor-interacting protein 13 | |
| Polyclonal | |
| Western Blot | |
| 16E1BP, 16E1-BP, HPV16 E1 protein binding protein, Human papillomavirus type 16 E1 protein-binding protein, thyroid hormone receptor interactor 13HPV16 E1 protein-binding protein, thyroid receptor interacting protein 13, thyroid receptor-interacting protein 13, TR-interacting protein 13, TRIP-13 | |
| The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human Thyroid Hormone Receptor Interactor 13 using the numbering given in entry NP_004228.1 (GeneID 9319). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 9319 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction