missing translation for 'onlineSavingsMsg'
Learn More

Thyroid receptor-interacting protein 12 Antibody - BSA Free, Novus Biologicals™

Product Code. 18322141 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322141 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322141 Supplier Novus Biologicals Supplier No. NBP281864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Thyroid receptor-interacting protein 12 Polyclonal specifically detects Thyroid receptor-interacting protein 12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroid receptor-interacting protein 12
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 2-20 ug/ml, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 6.3.2.-, KIAA0045ULF, MGC138849, MGC138850, probable E3 ubiquitin-protein ligase TRIP12, thyroid hormone receptor interactor 12, thyroid receptor interacting protein 12, Thyroid receptor-interacting protein 12, TR-interacting protein 12, TRIP-12
Gene Symbols TRIP12
Host Species Rabbit
Immunogen Thyroid receptor-interacting protein 12 antibody was raised against a 18 amino acid peptide near the carboxy terminus of human Thyroid receptor-interacting protein 12. The immunogen is located within amino acids 1930 - 1980 of Thyroid receptor-interacting protein 12. Amino Acid Squence: IVRKTFESTENPDDFLPS
Molecular Weight of Antigen 220 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9320
Test Specificity Thyroid receptor-interacting protein 12 antibody is human, mouse and rat reactive. At least four isoforms are known to exist.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.