missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid receptor-interacting protein 12 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £366.00
Specifications
| Antigen | Thyroid receptor-interacting protein 12 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679321
|
Novus Biologicals
NBP2-94325-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620122
|
Novus Biologicals
NBP2-94325-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thyroid receptor-interacting protein 12 Polyclonal antibody specifically detects Thyroid receptor-interacting protein 12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Thyroid receptor-interacting protein 12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 9320 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 6.3.2.-, KIAA0045ULF, MGC138849, MGC138850, probable E3 ubiquitin-protein ligase TRIP12, thyroid hormone receptor interactor 12, thyroid receptor interacting protein 12, Thyroid receptor-interacting protein 12, TR-interacting protein 12, TRIP-12 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human TRIP12 (NP_001271143.1). MSNRPNNNPGGSLRRSQRNTAGAQPQDDSIGGRSHLGQAKHKGYSPPESRKSNSKAPKVQSNTTSELSRGHLSKRSCSSSSAVIVPQPEDPDRANTSERQKTGQVPKKDNSRGVKRSASPDYNRTNSPSSAKKPKALQHTESPSETNKPHSKSKKRHLDQEQQLKSAQSPSTSKAHTRKSGATGGSRSQKRKRTESSCVKSGSGSESTGAEERSAKPTKLASKSATSAKAGCSTITDSSS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title