missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid receptor-interacting protein 12 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81864AF647
This item is not returnable.
View return policy
Description
Thyroid receptor-interacting protein 12 Polyclonal antibody specifically detects Thyroid receptor-interacting protein 12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Thyroid receptor-interacting protein 12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 647 | |
| EC 6.3.2.-, KIAA0045ULF, MGC138849, MGC138850, probable E3 ubiquitin-protein ligase TRIP12, thyroid hormone receptor interactor 12, thyroid receptor interacting protein 12, Thyroid receptor-interacting protein 12, TR-interacting protein 12, TRIP-12 | |
| Thyroid receptor-interacting protein 12 antibody was raised against a 18 amino acid peptide near the carboxy terminus of human Thyroid receptor-interacting protein 12. The immunogen is located within amino acids 1930 - 1980 of Thyroid receptor-interacting protein 12. Amino Acid Squence: IVRKTFESTENPDDFLPS | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 9320 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction