missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thyroid Peroxidase Recombinant Rabbit Monoclonal Antibody (JE60-40)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA550707

Product Code. 17919562

  • £414.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive control: Human thyroid tissue, human thyroid carcinoma tissue. Predicted band size: 103 kDa Subcellular Location: Membrane.

Thyroid Peroxidase is a gene that encodes for a membrane-bound glycoprotein, which functions as an enzyme and plays a crucial role in the proper functioning of the thyroid gland. This protein is responsible for the iodination of tyrosine residues in thyroglobulin and the formation of phenoxy-ester between pairs of iodinated tyrosines, which leads to the production of thyroid hormones such as thyroxine and triiodothyronine. Mutations in this gene are known to cause several disorders related to thyroid hormone production, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Several transcript variants encoding distinct isoforms have been identified for this gene, but the full-length nature of some variants remains undetermined. Diseases associated with TPO include Thyroid Dyshormonogenesis 2A and Congenital Hypothyroidism.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thyroid Peroxidase
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P07202
TPO
Recombinant protein within Human Thyroid Peroxidase/TPO aa 834-933/933.
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin)
JE60-40
Unconjugated
TPO
EC 1.11.1; LOW QUALITY PROTEIN: thyroid peroxidase; MSA; TDH2A; thyroid microsomal antigen; thyroid peroxidase; thyroid peroxidase (332 AA); thyroperoxidase; TPO; TPX; TPXEC 1.11.1.8
Rabbit
Protein A
RUO
7173
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Promociones

Promociones

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado