missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid Peroxidase Mouse anti-Human, HRP, Antibody Pair, Novus Biologicals™
Antibody pair for Solid phase sandwich ELISA
£285.00 - £587.00
Specifications
| Antigen | Thyroid Peroxidase |
|---|---|
| Applications | ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18330942
|
Novus Biologicals
NBP3-06708-5PLATES |
5 Plates |
£285.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18375693
|
Novus Biologicals
NBP3-06708-15PLATES |
15 Plates |
£587.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Solid phase sandwich ELISA for quantitative determination of Human Thyroid Peroxidase.Specifications
| Thyroid Peroxidase | |
| Monoclonal | |
| Mouse | |
| Lipid and Metabolism | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Storage of components varies. See protocol for specific instructions. |
| ELISA | |
| HRP | |
| RUO | |
| Human | |
| 7173 | |
| Antibody Pairs |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title