missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroid Peroxidase Antibody (TPO/7088R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21049JF635
This item is not returnable.
View return policy
Description
Thyroid Peroxidase Monoclonal antibody specifically detects Thyroid Peroxidase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Thyroid Peroxidase | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 | |
| Recombinant fragment of human Thyroid Peroxidase (around aa 600-900) (Exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 7173 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| TPO/7088R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur