missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Thyroglobulin Native Protein

Product Code. 18675786 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18675786

Brand: Novus Biologicals™ NBP262537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
For Use With (Application) ELISA
Formulation PBS
Gene ID (Entrez) 7038
Molecular Weight (g/mol) MolecularWeight-theroretical: 660 kDa
Quantity 1 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias AITD3TGN, TDH3, Tg, thyroglobulin
Gene Symbol TG
Research Category Cancer, Cell Biology
Purity or Quality Grade Size Exclusion Chromatography
Protein Thyroglobulin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.