missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Mouse, Clone: SPM221, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32904-0.1mg
This item is not returnable.
View return policy
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Thyroglobulin | |
| Monoclonal | |
| Unconjugated | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Thyroglobulin is a 660kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms. | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| SPM221 | |
| Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1.0ug/ml | |
| P01266 | |
| TG | |
| Human thyroid follicular cells | |
| 0.1 mg | |
| Cancer, Cell Biology | |
| 7038 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction