Learn More
Thyroglobulin Mouse, Clone: SPM221, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-32904-0.02MG
 View more versions of this product
Product Code. 18103635
  • £211.00 / 0.02mg
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thyroglobulin
Monoclonal
Unconjugated
PBS with 0.05% BSA. with 0.05% Sodium Azide
AITD3TGN, TDH3, Tg, thyroglobulin
Mouse
Protein A purified
RUO
Primary
Thyroglobulin is a 660kDa dimeric pre-protein with mutiple glycosylation sites. It is produced by and processed within the thyroid gland to produce the hormone thyroxine and triiodothyronine. Prior to forming dimers, thyroglobulin monomers undergo conformational maturation in the endoplasmic reticulation. The vast majority of follicular carcinomas of the thyroid will give positive immunoreactivity for anti-thyroglobulin even though sometimes only focally. Poorly differentiated carcinomas of the thyroid are frequently anti-thyroglobulin negative. Adenocarcinomas of other-than-thyroid origin do not react with this antibody. This antibody is useful in identification of thyroid carcinoma of the papillary and follicular types. Presence of thyroglobulin in metastatic lesions establishes the thyroid origin of tumor. Anti-thyroglobulin, combined with anti-calcitonin, can identify medullary carcinomas of the thyroid. Furthermore, anti-thyroglobulin, combined with anti-TTF1, can be a reliable marker to differentiate between primary thyroid and lung neoplasms.
Store at 4C.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
SPM221
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1.0ug/ml
P01266
TG
Human thyroid follicular cells
0.02 mg
Cancer, Cell Biology
7038
Human, Mouse, Rat
Purified