missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Mouse anti-Human, Mouse, Rat, Clone: rTGB/4744, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Thyroglobulin |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rTGB/4744 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 0.5-1 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | AITD3TGN, TDH3, Tg, thyroglobulin |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?