missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thyroglobulin Monoclonal Antibody (TG60), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

£367.00 - £916.00

Specifications

Antigen Thyroglobulin
Clone TG60
Concentration 2.0 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17820176
View Documents
Invitrogen™
MA5-46102
100 μg
£367.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11538882
GREENER_CHOICE
View Documents
Invitrogen™
MA110593
500 μg
£439.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17890476
View Documents
Invitrogen™
MA5-45989
1 mg
£916.00
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MA1-10593 detects Thyroglobulin, specifically both T3 and T4, from human samples. MA1-10593 has been successfully used in ELISA applications. The MA1-10593 immunogen is purified human Thyroglobulin.

The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
Specifications

Specifications

Thyroglobulin
2.0 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor
TG
IgG1
Protein A
Antibody
TG60
ELISA
Unconjugated
Mouse
Human
P01266
7038
Purified human Thyroglobulin.
Primary
-20°C, Avoid Freeze/Thaw Cycles
TG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.